Would you like to log in again?
VWR® 3500I Incubating Orbital Shaker
Supplier: VWR
VWR®
80863468 80863469
12620-946EA 7249.6 USD InStock
12620-946 10027-242
VWR® 3500I Incubating Orbital Shaker
Shakers and Mixers Shaking Incubators
The microprocessor control of the Model 3500I provides consistent, uniform mixing action.
  • Microprocessor control with digital display for speed, temperature, and time
  • Opti-flow forced ventilation system
  • Triple eccentric drive with brushless motor
  • Five-year warranty
  • NIST-Traceable, ISO/IEC 17025 accredited calibration certificate from Troemner available
A spill-resistant touchpad adjusts the temperature in 1 °C increments
For added safety, a ramping feature increases speed to set point, and a built-in load sensor automatically reduces rpm during unbalanced conditions to further prevent spills and protect samples. The brushless motor is designed to handle heavy load, and the spill-resistant design protects internal components from accidental spills and leaks.
The timer displays elapsed time or counts to zero and shuts off the unit when programmed to user-defined limit. An alarm sounds when time reaches zero. The temperature-calibration mode allows the unit to be calibrated to an external temperature device, and the Opti-Flow forced ventilation system uses twin induction fans and air deflectors to deliver temperature uniformity and stability. The Accu-Drive™ Shaking System ensures precise speed control. This system monitors shaking speed and maintains set point even under changing loads.
Certifications: RoHS certified. TUV Listed. Calibrated shakers include a Troemner ISO/IEC accredited calibration certificate traceable to NIST. The ISO/IEC 17025 accredited calibration certificate includes multiple data points within the temperature and speed ranges and time function with the associated uncertainties. Troemner is accredited by NVLAP under laboratory code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: This model includes a nonskid mat for the tray and a 2.3 m, 7.6', detachable, three-wire cord and plug. 230 V, 50/60 Hz models are also available. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty