Would you like to log in again?
VISOR UVEX TURBOSHIELD W/HARDCOAT PC CLR
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Uvex Turboshield* PC Visor^ Uvex Turboshield* PC Visor, color: clear, material: Polycarbonate, Providing Excellent Optics, Increased Field of View and Added Chin Coverage Without Bulky Attachment With Anti-Fog/Hardcoat, Developed through extensive customer research and featuring patented technology
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VISOR UVEX TURBOSHIELD W/HARDCOAT PC CLR 10027-351 S9555 Case of 10 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty