Would you like to log in again?
VWR® CryoPro® Rack Systems, BR Series Promotion Packages
Supplier: Avantor
VWR®
BR-3-PROMO14 BR-1-PROMO14 BR-2-PROMO14 BR-4-PROMO14
10027-530EA 9437.13 USD InStock
10027-530 10027-588 10027-532 10027-590
VWR® CryoPro® Rack Systems, BR Series Promotion Packages
Cryopreservation Systems
Combining the convenience of box vial storage with the economy of a low nitrogen loss design, the BR Series provides the perfect solution for medium to large storage needs.
  • Convenient box vial storage
  • Low nitrogen loss
Vial storage in boxes allows for quick and accurate inventorying and recovery of specimens. The BR-1 model can hold 25-cell cardboard or plastic boxes. The BR-2, BR-3, and BR-4 models can hold 100-cell cardboard or plastic boxes.
Ordering information: Supplied with rollerbase, full capacity of fibreboard storage boxes and 1 pair of cryogenic gloves.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty