Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clinical-Grade Centrifuge Tube, 15 ml, Globe Scientific

Supplier: Globe Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

6287 6289 6285 6284 6288 6286
76452-258CS 201.8 USD InStock
76452-258 76452-256 10027-868 10027-866 89209-794 76452-260
Clinical-Grade Centrifuge Tube, 15 ml, Globe Scientific
Tubes Centrifuge Tubes
These disposable conical-bottom centrifuge tubes have been specially designed for clinical applications.

  • Available in 15 and 50 ml versions
  • Easy-to-read black graduation marks in 0.5 ml increments
  • White writing area for sample identification

The blue, high-density polyethylene screw caps ensure a leak-resistant and tight seal. Produced from a special grade of polypropylene, these tubes offer exceptional clarity and chemical stability.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty