Would you like to log in again?
Low-Profile Glove Dispensers, S-Curve
Supplier: S-CURVE TECHNOLOGIES
AT-105 PETG AT-105
10027-870EA 716.91 USD InStock
10027-870 10027-756
Low-Profile Glove Dispensers, S-Curve
Glove Dispensers and Glove Box Holders
Low profile wall mount glove dispenser with front access holes. Designed to hold multiple glove sizes in a low-profile dispenser.
  • Sloped top to maintain HEPA airflow
  • Full-length hinge for durability
  • Mounts to wall
  • Wall bracket included
  • Flame treated edges for a smooth finish
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty