Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Ser-NHNH₂ ≥98% (by HPLC), powder

Supplier: Avantor

0424425GM
100271-432EA 194 USD InStock
100271-432
Z-Ser-NHNH₂ ≥98% (by HPLC), powder
Z-Ser-NHNH₂
  • White powder
  • Store at 0-8 °C


Formula: C₁₁H₁₅N₃O₄
Melting Pt: 182…186 °C
MDL Number: MFCD00153341
CAS Number: 26582-86-5
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/24/21286024.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/60/25/21286025.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty