Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-α-Hydroxybenzoic acid ≥98%, solid

Supplier: Avantor

14220250MG
100274-862EA 441 USD InStock
100274-862
DL-α-Hydroxybenzoic acid ≥98%, solid
DL-α-Hydroxybenzoic acid
  • Solid
  • Store in a cool, dry place


Formula: C₂₂H₄₄O₃
MDL Number: MFCD00083370
CAS Number: 13980-14-8
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/62/04/21286204.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty