Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-hydroxydocosanoate ≥98%

Supplier: Avantor

24220250MG
100275-022EA 418 USD InStock
100275-022
Methyl-2-hydroxydocosanoate ≥98%
Methyl-2-hydroxydocosanoate
Store in a cool, dry place


Formula: C₂₃H₄₆O₃
MDL Number: MFCD00083372
CAS Number: 13980-17-1
ADR: ,
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty