Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Proline ethyl ester hydrochloride ≥97%, powder

Supplier: Avantor
Synonyms: H-D-Pro-OEt hydrochloride

0417525GM
100279-734EA 135 USD InStock
100279-734
D-Proline ethyl ester hydrochloride ≥97%, powder
D-Proline ethyl ester hydrochloride
  • White powder
  • Store at 0-8 °C


Formula: C₇H₁₄ClNO₂
MW: 179.644 g/mol
MDL Number: MFCD00191003
CAS Number: 131477-20-8
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/73/51/21287351.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/73/52/21287352.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty