Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride ≥100%, powder

Supplier: Avantor
Synonyms: 4-trans-Hydroxy-L-proline ethyl ester hydrochlorideH-Hyp-OEt·HCl(2S,4R)-Ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride

0418225GM
100279-816EA 41 USD InStock
100279-816
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride ≥100%, powder
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride
  • White powder
  • Store at 0-8 °C


Formula: C₇H₁₃NO₃·HCl
MW: 195.643 g/mol
Melting Pt: 135…138 °C
MDL Number: MFCD00040589
CAS Number: 33996-30-4
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/65/91/21286591.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/65/90/21286590.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty