Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunclon™ Sphera™ Microplates, Dishes, and Flasks, Thermo Scientific
Supplier: Thermo Scientific
Nunclon™
174930 174952 174929 174927 174932 174925 174951 174945 174931 174944 174943
10028-032CS 432.12 USD InStock
10028-032 10028-020 10028-034 10028-022 10028-036 10028-024 10028-038 10028-026 76323-842 10028-028 10028-030
Nunclon™ Sphera™ Microplates, Dishes, and Flasks, Thermo Scientific
Culture Dishes Cell Culture Dishes
The Thermo Scientific™ Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment
  • Optimises spheroid formation
  • Supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth
  • Promotes formation of embryoid bodies (EB)
The Nunclon™ Sphera™ surface minimizes variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumor growth, providing a better model system for studying tumor cell progression and efficacy of anticancer agents in vitro.
The new surface supports many different cell types and their ability to generate spheroids; studies show that over time the spheroids grow in volume, indicating that the new surface enables consistent, repeatable cell growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty