Would you like to log in again?
CurTec Polypropylene Lids for Packo Containers, Qorpak®
Supplier: Qorpak
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
242936 242941 243030 248746 242938 248747 242937 248749 242943 248748 243028 248750
10028-660CS 54.46 USD InStock
10028-660 10028-652 76339-910 10028-654 76339-912 10028-656 76339-914 10028-658 76339-904 76339-916 76339-906 76339-908
CurTec Polypropylene Lids for Packo Containers, Qorpak®
Closures Bottle Closures
Packo Lids are available with an integrated gasket, or plain for use with inner seals also (sold separately). This High Performance Packaging line provides the highest level of product protection and added value through quality, functionality, ease of use, and design.
  • Quarter-turn screw lid
  • Water vapor tight
  • Integrated tamper evident strip
  • UN certified with Packo jars
Suggested for use with pharmaceuticals, fine chemicals, food ingredients, and hazardous waste. Airtight design prevents moisture contamination.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty