Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Natural Air Circulation Incubators Units, Memmert

Supplier: Memmert

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

IN75, 115 V IN110PLUS115 IN450115V IN30, 230 V IN75PLUS115 IN160, 115 V IN260, 230 V IN55, 230V IN450PLUS115 IN75, 230 V IN750PLUS115 IN450PLUS230 IN450230V IN750PLUS230 IN30, 115 V IN260PLUS115 IN110PLUS230 IN110, 115 V IN55, 115 V IN55PLUS115 IN30PLUS115 IN30PLUS230 IN160, 230 V IN55PLUS230 IN160PLUS115 IN260PLUS230 IN750230V IN75PLUS230 IN110, 230 V IN260, 115 V IN750115V IN160PLUS230
10861-402EA 14163.27 USD InStock
10861-402 10028-970 10861-388 10028-950 10028-972 10861-400 10028-942 10028-964 10028-944 10028-966 10028-946 10028-968 10028-948 10028-960 10861-398 10028-962 10028-940 10861-394 10861-396 10861-390 10861-392 10028-938 10028-930 10028-974 10028-952 10028-932 10028-954 10028-976 10028-934 10028-956 10028-958 10028-936
Natural Air Circulation Incubators Units, Memmert
Incubators Standard Incubators
The Generation 2012 range has natural and forced convection models with a temperature range up to +80 °C

SingleDISPLAY models; ideal for standard applications. ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, programme time, time zones and daylight-saving time. One PT100 temperature sensor in a 4-wire circuit and ethernet interface for reading out the protocol log. Double over-temperature protection including visual alarm: Electronic temperature monitoring with freely adjustable monitoring temperature and mechanical temperature limiter TB acc. to DIN 12 880 which switches off the heating if the maximum permitted temperature is exceeded by approx. 10 °C, protecting the oven.

TwinDISPLAY models; ideal for complex ramping processes. Intuitive, user friendly ovens with 2 TFT displays. ControlCOCKPIT with USB port. ControlCOCKPIT parameters as above. Two PT100 sensors in a 4-wire circuit for mutual monitoring, taking over functions in case of an error. The HeatBALANCE function allows application specific adjustment of heating power distribution (balance) between the upper and lower heating areas. Logged protocol data is displayed on the ControlCOCKPIT (max 10,000 values correspond to approx. 1 week). Ethernet interface for reading out the protocol log and for uploading and implementing programmes and for online logging. Multiple overtemperature protection with audible and visual alarm: Electronic temperature monitoring TWW/TWB adjustable on display (independent adjustable temperature safety device class 3.1, which protects the samples and the oven) and mechanical temperature limiter TB (protection class 1) acc. to. DIN 12 880. AutoSAFETY automatically adjusts to the set value within a freely adjustable tolerance range. Setting individual MIN/MAX values for over-/under-temperature.

There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application. All models have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. These incubators have an integrated data logger and digital timer which is adjustable from 1 minute to 99 days and 23 hours. All models with TwinDISPLAY feature the intuitive control and logging software, AtmoCONTROL, which uses drag and drop symbols to input values. The software allows online monitoring of up to 32 connected appliances and can send an automatic alarm message to one or several e-mail addresses. INplus and IFplus units have a 4 hour sterilisation cycle at 160 °C. Complex ramping processes can be programmed quickly and easy for TwinDISPLAY models. The SetpointWAIT function ensures the process time does not start before the set temperature is reached. This feature can also be used with additional, freely positionable PT100 sensors, so that the process time starts when the set temperature is reached at all measuring points. Stainless Steel color.

Certifications: CE Marked. UL listed.

Ordering information: Range of accessories and factory fitted options such as access ports, interior sockets, lighting and interior door are available on request. Please contact Avantor for further details prior to ordering as these can not be retro-fitted.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty