Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Natural Air Circulation Ovens Units

Supplier: Memmert

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

UN260PLUS115 UN30, 230 V UN55, 115 V UN110PLUS115 UN450PLUS3PH UN30PLUS 230 UN160PLUS115 UN160, 115 V UN110, 230 V UN260, 115 V UN75PLUS,115 UN110PLUS230 UN260, 230 V UN55PLUS,230 UN260PLUS230 UN75, 115 V UN30, 115 V UN30PLUS 115 UN75PLUS,230 UN55PLUS,115 UN160PLUS230 UN55, 230 V UN110, 115 V UN160, 230 V UN75, 230 V UN450230V3PH
10028-990EA 4897.64 USD InStock
10028-990 10861-404 10029-018 10028-992 10029-016 10028-994 10028-908 10029-004 10028-986 10029-002 10028-988 10029-022 10029-000 10029-020 10028-980 10028-982 10029-008 10028-984 10029-006 10028-996 10029-014 10029-012 10028-998 10028-978 10861-406 10029-010
Natural Air Circulation Ovens Units
Ovens Standard Ovens
The Generation 2012 range has natural and forced convection models with a temperature range up to +300 °C.

SingleDISPLAY models; ideal for standard applications. ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, programme time, time zones and daylight-saving time. One PT100 temperature sensor in a 4-wire circuit and ethernet interface for reading out the protocol log. Double over-temperature protection including visual alarm: Electronic temperature monitoring with freely adjustable monitoring temperature and mechanical temperature limiter TB acc. to DIN 12 880 which switches off the heating if the maximum permitted temperature is exceeded by approx. 10 °C, protecting the oven.

TwinDISPLAY models; ideal for complex ramping processes. Intuitive, user friendly ovens with 2 TFT displays. ControlCOCKPIT with USB port. ControlCOCKPIT parameters as above. Two PT100 sensors in a 4-wire circuit for mutual monitoring, taking over functions in case of an error. The HeatBALANCE function allows application specific adjustment of heating power distribution (balance) between the upper and lower heating areas. Logged protocol data is displayed on the ControlCOCKPIT (max 10000 values correspond to approx. 1 week). Ethernet interface for reading out the protocol log and for uploading and implementing programmes and for online logging. Multiple overtemperature protection with audible and visual alarm: Electronic temperature monitoring TWW/TWB adjustable on display (independent adjustable temperature safety device class 3.1, which protects the samples and the oven) and mechanical temperature limiter TB (protection class 1) acc. to. DIN 12 880. AutoSAFETY automatically adjusts to the set value within a freely adjustable tolerance range. Setting individual MIN/MAX values for over-/under-temperature.

There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application. All models have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. These ovens have an integrated data logger and digital timer which is adjustable from 1 minute to 99 days and 23 hours. All models with TwinDISPLAY feature the intuitive control and logging software, AtmoCONTROL, which uses drag and drop symbols to input values. The software allows online monitoring of up to 32 connected appliances and can send an automatic alarm message to one or several e-mail addresses. Complex ramping processes can be programmed quickly and easily for TwinDISPLAY models. The SetpointWAIT function ensures the process time does not start before the set temperature is reached. This feature can also be used with additional, freely positionable PT100 sensors, so that the process time starts when the set temperature is reached at all measuring points. Stainless Steel color.

Certifications: CE Marked. UL listed.

Ordering information: Range of accessories and factory fitted options such as access ports, interior sockets, lighting and interior door are available on request. Please contact Avantor for further details prior to ordering as these can not be retro-fitted. Please note U 450 and U 750 units require a 208 V three phase electrical supply.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty