Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nɑ-(tert-Butoxycarbonyl)-L-tryptophan N-succinimidyl ester ≥98% (by HPLC), powder

Supplier: Avantor
Synonyms: Boc-Trp-OSuNɑ-Boc-L-tryptophan N-Succinimidyl Ester

049125GM
100281-996EA 102 USD InStock
100281-996
Nɑ-(tert-Butoxycarbonyl)-L-tryptophan N-succinimidyl ester ≥98% (by HPLC), powder
Nalpha-(tert-Butoxycarbonyl)-L-tryptophan N-succinimidyl ester
  • White to off-white powder
  • Store at 0-8 °C


Formula: C₂₀H₂₃N₃O₆
MW: 401.42 g/mol
Melting Pt: 141…146 °C
MDL Number: MFCD00022756
CAS Number: 3392-11-8
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/67/80/21286780.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/67/79/21286779.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty