Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Carboxy-2-chloro-a,a,a-trifluoroanisole ≥97%

Supplier: Avantor
Synonyms: 3-Chloro-4-(trifluoromethoxy)benzoic acid

072525GM
100282-376EA 118 USD InStock
100282-376
4-Carboxy-2-chloro-a,a,a-trifluoroanisole ≥97%
4-Carboxy-2-chloro-a,a,a-trifluoroanisole


Formula: C₈H₄ClF₃O₃
MW: 240.56 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD01631556
CAS Number: 158580-93-9
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/68/02/21286802.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty