Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6(Z),9(Z),12(Z),15(Z),18(Z)-Heneicosapentaenoic Acid

Supplier: Avantor

10210515MG
100284-020EA 688 USD InStock
100284-020
6(Z),9(Z),12(Z),15(Z),18(Z)-Heneicosapentaenoic Acid
6(Z),9(Z),12(Z),15(Z),18(Z)-Heneicosapentaenoic acid


Formula: C₂₁H₃₂O₂
MDL Number: MFCD05864037
CAS Number: 24257-10-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty