Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-hydroxyoctanoate ≥98%

Supplier: Avantor

24080250MG
100285-396EA 409 USD InStock
100285-396
Methyl-2-hydroxyoctanoate ≥98%
Methyl-2-hydroxyoctanoate
Store in a cool, dry place


Formula: C₉H₁₈O₃
MDL Number: MFCD00673452
CAS Number: 92572-96-8
ADR: ,
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty