Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromoperfluorohexane ≥98%

Supplier: Avantor
Synonyms: Perfluorohexyl bromide

270525GM
100289-334EA 85 USD InStock
100289-334
1-Bromoperfluorohexane ≥98%
1-Bromoperfluorohexane


Formula: BrF₂C(CF₂)₄CF₃
MW: 398.95 g/mol
Boiling Pt: 100…101 °C
MDL Number: MFCD00042349
CAS Number: 335-56-8
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/47/21287047.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty