Would you like to log in again?
Shipper, Category B Refrigerated, 95 kPa Bags, 18-Tube, Therapak®
Supplier: Therapak
37816
10029-300CS 190.62 USD InStock
10029-300
Shipper, Category B Refrigerated, 95 kPa Bags, 18-Tube, Therapak®
Mailers and Shippers
Refrigerated Shipping System includes transport box (OD: 15x13x12"), medium cooler (ID: 12x10x9"), three 95kPa bags, three Aqui-Pak™ segmented absorbent pouches, 8 oz
refrigerant packs, cushioning material, list of contents card, and required labels for IATA PI650 marking and compliance. Holds up to eighteen tubes for overnight shipments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty