Would you like to log in again?
Single Specimen Collection Kits, Therapak®
Supplier: Therapak
40010 40012G 40009 40003 40004 40005
89428-908CS 227.5 USD InStock
89428-908 89171-052 10029-338 10029-344 10029-342 10029-340
Single Specimen Collection Kits, Therapak®
Sample Containers General Purpose Containers
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis.
Each kit contains a leak-resistant 90 ml sterile container with cap and integrity seal. The transport bag features two sealable, tamper-evident pockets, and comes with absorbent material. The individually bagged kits are available in various configurations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty