Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Split Specimen Collection Kits, Therapak®
Supplier: Therapak
40011 40002
10029-336CS 326.84 USD InStock
10029-336 10029-346
Split Specimen Collection Kits, Therapak®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
For drugs of abuse testing applications requiring two samples to be collected from a single donor, the split specimen collection and transport kits are available in either a split bag kit or the same kit within a transport box.
Delivery information: Kits include 6.5 oz. beaker style collection cup with pour-spout and temperature strip, 2 - 60 mL sterile containers with sterility seal and a transport bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty