Would you like to log in again?
Shipper, NanoCool, Refrigerated 2...8 °C, Therapak®
Supplier: Therapak
56849 56850
10029-390CS 520.12 USD InStock
10029-390 10029-388
Shipper, NanoCool, Refrigerated 2...8 °C, Therapak®
Mailers and Shippers
Therapak®’s NanoCool 2-8°C Specimen Shipping System features a self-cooling system built into the lid.
Temperature performance testing shows the system maintains 2-8°C when tested against a ISTA-7D 48-hour summer profile. The NanoCool’s durable design also passes drop testing to a modified ISTA-3A sequence.
With no refrigeration required, activate the cooling process with the simple push of a button and your contents will remain in the 2° to 8°C range for up to 48 hours with no further conditioning or temperature control necessary.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty