Would you like to log in again?
Shipper, EAS Ambient, Small Transport Box, Therapak®
Supplier: Therapak
56880
10029-410CS 211.56 USD InStock
10029-410
Shipper, EAS Ambient, Small Transport Box, Therapak®
Mailers and Shippers
The small transport box is designed for shipping one standard or larger specimen container (30 mL to 60 mL) of liquid specimen.
Shipped with pre-assembled boxes, these transport shippers also include: small transport box (ID: 5.25 x 4 x 3.75"; OD: 6 x 4.25 x 4"), specimen transport bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label and cushioning material where applicable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty