Would you like to log in again?
Shipper, EAS Refrigerated, Therapak®
Supplier: Therapak
56884
10029-418CS 90.82 USD InStock
10029-418
Shipper, EAS Refrigerated, Therapak®
Mailers and Shippers
EAS shipping systems for refrigerated specimens feature Therapak®'s standard small insulated cooler (ID: 8 x 6 x 8”) and transport box (OD: 11 x 9 x 11”), which is ideal for overnight shipments via common carriers such as DHL and FedEx
Refrigerated shipping system includes: 8-oz cold packs, specimen bag, absorbent material, Exempt Animal Specimen label, biohazard label and cushioning material. Each case includes one set of packing instructions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty