Would you like to log in again?
Aldehyde-Ketone-DNPH TO-11A Calibration Mix (15 components), Restek
Supplier: Restek
31808
10029-522EA 106.74 USD InStock
10029-522
Aldehyde-Ketone-DNPH TO-11A Calibration Mix (15 components), Restek
Standards Organic Standards
This consists of acetaldehyde-DNPH; acetone-DNPH; acrolein-DNPH; benzaldehyde-DNPH; n-butyraldehyde-DNPH; crotonaldehyde-DNPH; 2,5-dimethylbenzaldehyde-DNPH; formaldehyde-DNPH; hexaldehyde-DNPH; isovaleraldehyde-DNPH; propionaldehyde-DNPH; m-tolualdehyde-DNPH; o-Tolualdehyde-DNPH; p-tolualdehyde-DNPH and aleraldehyde-DNPH.
Caution: *The reported concentrations reflect the amount of aldehyde or ketone in the mixture. The concentration of derivatized aldehyde or ketone is not reported.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty