Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetylsphingosylphosphorylcholine ≥98%

Supplier: Avantor

5610825MG
100291-334EA 755 USD InStock
100291-334
N-Acetylsphingosylphosphorylcholine ≥98%
N-Acetylsphingosylphosphorylcholine
Store in a cool, dry place


MDL Number: MFCD02259129
ADR: ,
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty