Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Leu-Arg-Pro-Oh

Supplier: Avantor

042002250MG
100296-598EA 1632.75 USD InStock
100296-598
H-Leu-Arg-Pro-Oh
H-Leu-Arg-Pro-Oh


Formula: C₁₇H₃₂N₆O₄
MDL Number: MFCD05662180
CAS Number: 133943-59-6
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/43/21287443.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty