Would you like to log in again?
Integra® EC, Extended Cuff Blue Nitrile Gloves, Microflex®
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
N871 N872 N874 N874 N875 N873 N872 N875 N871 N873
10030-250BX 48.22 USD InStock
10030-250 CA10030-248 10030-246 CA10030-246 10030-248 CA10030-252 10030-242 CA10030-242 10030-252 CA10030-250
Integra® EC, Extended Cuff Blue Nitrile Gloves, Microflex®
Gloves
Integra EC is a durable, 8-millimeter glove for tough jobs where extra durability or safety is needed.
  • Ambidextrous
  • Non sterile
  • Beaded cuffs
  • Durable and extra thick
The added thickness of these gloves gives strength and durability. Great protection against bloodborne pathogens. Extra thickness and extended cuff provide extra protection. Gloves are blue, nonsterile, and ambidextrous. They feature a beaded cuff and are fully textured.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty