Would you like to log in again?
Biopsy Cassettes, FastLoad Taped Stacks, Closed Lid, Azer Scientific
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ES6016-T ES6016-W ES6016-A ES6016-GY ES6016-P ES6016-G ES6016-B ES6016-L ES6016-Y ES6016-O ES6016-PH
10030-460CS 770.69 USD InStock
10030-460 10030-470 10030-458 10030-466 10030-454 10030-468 10030-456 10030-462 10030-472 10030-464 10030-474
Biopsy Cassettes, FastLoad Taped Stacks, Closed Lid, Azer Scientific
Embedding Cassettes Biopsy Processing Cassettes
FastLoad Biopsy Taped Cassettes feature a recessed hinged lid that can be opened and closed as often as necessary and always relocks securely.
  • Attached lid
  • Cassettes taped for automated systems
  • Histology-solvents resistant
  • Three frosted writing areas
The lid is attached and in the closed position.
Cassettes are molded from a high-density polymer.
Packaging: Cassettes are packaged in taped stacks of 40. Each case contains 50 stacks for a total of 2000 per case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty