Would you like to log in again?
Tissue/Biopsy Cassettes, FastLoad Taped Stacks, Open Style, Azer Scientific
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ES6013-O ES6012-O ES6013-T ES6012-B ES6013-B ES6013-G ES6012-Y ES6013-P ES6013-Y ES6012-L ES6013-GY ES6012-P ES6012-GY ES6012-W ES6012-PH ES6013-W ES6013-L ES6012-G ES6013-A ES6013-PH
10030-518CS 369.92 USD InStock
10030-518 10030-408 10030-506 10030-508 10030-404 10030-514 10030-502 10030-406 10030-516 10030-504 10030-400 10030-488 10030-510 10030-512 10030-402 10030-500 10030-494 10030-398 10030-496 10030-492
Tissue/Biopsy Cassettes, FastLoad Taped Stacks, Open Style, Azer Scientific
Embedding Cassettes Biopsy Processing Cassettes
Tissue and Biopsy Cassettes in FastLoad Taped Stacks are constructed from a special, high-density polymer that is resistant to histology solvents.
  • One-piece design
  • Open-position attached lids
  • Anterior writing area at 35° angle
One piece snap-latch and hinge lock design allows one hand operation. The lids are attached in an open position for easy filling but can be opened and closed as often as necessary. The anterior writing area is at a 35° angle.
Packaging: Packaged in stacks of 40. Each case contains 25 stacks for a total of 1000 casettes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty