Would you like to log in again?
Palletech® Handwrap, Associated Bag
Supplier: Associated Bag
20-20
10030-708CT 191.16 USD InStock
10030-708
Palletech® Handwrap, Associated Bag
Tapes
Palletech® Handwrap retains strength and cling in cold temperatures.
  • Resistant to Cold Temperature Effects
  • Three-Inch Cores
  • Top-Grade Resin
  • Superior Stretch and Cling
  • Puncture-Resistant
Top-grade resin produces superior stretch, cling, puncture-resistance, and clarity.
Rolls wraps and stabilize light to medium loads. They have three-inch cores.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty