Would you like to log in again?
Low Density Flat Poly Bags, Associated Bag
Supplier: Associated Bag
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
28-6-624 28-6-610 28-6-606 28-6-695 28-6-669 28-6-646 28-6-769 28-4-698 28-6-635 28-6-799 28-4-514 28-5-536 28-4-566 28-6-691 28-5-554 28-6-968 28-4-639 28-6-850 28-6-697 28-5-547 28-5-557
47743-777CS 239.96 USD InStock
47743-777 80086-340 47735-418 47743-779 80087-528 80086-342 80087-526 10030-712 47743-773 80086-344 47743-775 80086-346 47735-416 10030-710 47745-020 47746-468 11215-930 80086-338 47735-422 80086-348 47735-420
Low Density Flat Poly Bags, Associated Bag
Bags Storage Bags
Economical and reusable.
2 to 8 mil for light- to super-heavy weight uses. Not recommended for liquids.
Certifications: Meets FDA and USDA specifications for food contact.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty