Would you like to log in again?
ClipTip® 384 Pipette Tips, 125 μl, Thermo Scientific
Supplier: Thermo Scientific
ClipTip™
94410150 94410153 94420153
10030-810CS 1116.34 USD InStock
10030-810 10030-812 10030-808
ClipTip® 384 Pipette Tips, 125 μl, Thermo Scientific
Pipette Tips
Thermo Scientific ClipTip® 384 pipette tips feature a unique ‘Snap and Lip’ seal technology to attach and seal the tips on each channel with minimal tip attachment and ejection force.
  • Certified free of RNase, DNase, DNA, ATP, and pyrogen contamination
  • Each tip insert has ink-jetted lot and volume information
ClipTip® 384 model selection is optimized for 384-microplate and molecular biology applications. The tips are available in three sizes and packaging options, with 12.5, 30, and 125 μl tips available in non sterile, sterile, and filtered formats. A convenient color key makes it easy to match the color of the E1-ClipTip® pipettor with the corresponding ClipTip® pipette tip rack insert.
ClipTip® 384 125 µl pipette tips suit pipetting of 1 to 125 µl volumes with the E1-ClipTip® electronic pipettors.
Certifications: Manufactured according to ISO 9001, ISO 14001, and ISO 13485.
Ordering information: A lot specific Certificate of Conformity is provided with each unit box.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty