Would you like to log in again?
EvoControl 400 Cleanroom Wipes, Vileda Professional-FHP
Supplier: Vileda Professional
142856 142857
10030-968CS 3852.16 USD InStock
10030-968 10030-966
EvoControl 400 Cleanroom Wipes, Vileda Professional-FHP
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Maximize contamination control with Vileda® wipes! The microfiber EvoControl cleanroom wipes fulfill the highest demands for cleanroom wiping, plus EvoControl provides revolutionary wiping results thanks to its unique microfiber structure.
  • EvoControl 400 is a disposable liquid cleaning wipe designed for contamination removal
  • It is pre-laundered, laser cut and designed for the removal of acids and chemical spills
Evolon microfiber cleans glass and stainless steel without leaving residues or streaking and also has a low particle release while in use. The EvoControl cleanroom microfiber is 10 times finer than standard knitted PES cleanroom wipes, removing contamination more thoroughly along with maximizing particle pick up and bacteria removal. The cloths laser cut sealed edge are melted together for a perfect clean edge, which prevents particle release.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty