Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Beakers, Square Ratio, Polypropylene, Dynalon
Supplier: Dynalon
522085-0600 522085-0250 522085-1000 522085-0400 522085-2000 522085-0500 522085-1000 522085-0025 522085-0400 522085-0100 522085-0050
30620-140CS 92.08 USD InStock
30620-140 490012-454 10031-468 10031-470 470188-907 30617-170 30620-136 30620-146 30620-144 30621-000 30620-142
Beakers, Square Ratio, Polypropylene, Dynalon
Beakers
Square ratio beakers are a safe, convenient alternative to griffin style beakers
These beakers feature a vertical lip around the top of the beaker which serves two purposes: first the lip contains many spills that may occur with the rounded design of a griffin style beaker. Second, this vertical lip allows the beakers to be stacked together in sets when not in use. Maximum use temperature is 121°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty