Would you like to log in again?
PIG® Surgical Floor Skimmer
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HC200 HC201
10031-716BX 362.5 USD InStock
10031-716 10031-718
PIG® Surgical Floor Skimmer
Spill Control Materials
Revolutionary design attacks puddle problems from all sides to help keep O.R. floors dry and safe.
  • Cloverleaf shape directs liquids into the suction channel for maximum efficiency and performance
  • Proprietary bottom wicking pad removes 60 liters per hour
  • Top fluid collection channel captures liquid from the drapes and channels it to the wicking pad
  • Composition: Polypropylene/Cellulose Blend
  • Thin Low-profile design
Easy-slide floor pad can be repositioned or swept across the floor with your foot and captures liquids no matter where they go. Silent suction design helps keep the operating room quite. Suction tubing (included) allows access to all areas around the O.R. table.
Patent-pending design.
Accessories information: Latex-free suction tubing 3.66 mm.
Ordering information: Sold as 10 or 20 per box.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty