Would you like to log in again?
VWR® Workstation Organization
Supplier: VWR
15579 15578 15577
10031-838EA 394.25 USD InStock
10031-838 10031-836 10031-840
VWR® Workstation Organization
Racks Bottle Racks
Utilize vertical space of your benchtop with the Workstation Riser.
  • Chemically resistant polycarbonate construction eliminates spill worries
  • Clear fabrication provides strength and visibility
  • Easy organization of contents
Put all of your most important tools or frequently used products on the top shelf.
Manufactured with clear 0.5" acrylic (Benchtop Risers) for strength and visibility.
With the Workstation Riser products, take organization to the next level.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty