Would you like to log in again?
VWR® Side Load Glove Dispensers
Supplier: VWR
15692 15702 15671 15701 15700
10031-900EA 199.56 USD InStock
10031-900 10031-898 10031-892 10031-896 10031-894
VWR® Side Load Glove Dispensers
Glove Dispensers and Glove Box Holders
Unique Side-Loading Glove Dispensers have molded dividers that are incredibly strong and keep each box separated from the other.
  • Side-loading design
  • Strong molded dividers
  • Easy to use
This design removes the bothersome nature of reloading glove dispensers at the bottom of a stack. No need to remove other boxes during exchange. Boxes slide independently from dispenser side. They make switching out a fresh box for an empty one simple and fast so that you can attend to more important tasks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty