Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ecosense pH10A Handheld pH/Temperature Pen Tester, YSI

Supplier: YSI

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PH10
10032-016EA 196.51 USD InStock
10032-016
Ecosense pH10A Handheld pH/Temperature Pen Tester, YSI
Electrochemical Meters pH and Multi-Parameter Meters
The pH10A is part of the EcoSense® family of products. It's the perfect instrument for economical spot sampling of pH and temperature in many applications including wastewater, surface water, aquaculture, hydroponics, pools and education.

  • Waterproof IP 67
  • Automatic calibration and buffer recognition
  • 50-set memory (pH, temp, date and time stamp)
  • Replaceable single- or double-junction electrode
  • GLP functionality
  • 1-, 2- or 3-point calibration
  • Hold feature locks stable readings on the display
  • >200 h battery life with low battery indicator
  • Probe: Thermistor, 10kΩ / 25 °C

Certifications: CE compliance. IP-67 Waterproof Rating.

Ordering information: Instrument includes pH electrode, batteries and instruction manual. Replacement electrodes and batteries are sold seperately. Instrument comes with a 1-year instrument and 6-month electrode warranty. The pH10A (gray) directly replaces the pH10 (blue).

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty