Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Lab Coats, ESD Knit
Supplier: CritiCore Protective Wear
10032-314 10032-770 10032-308 10032-310 10032-312 10032-394 10032-768
10032-314EA 60.92 USD InStock
10032-314 10032-768 10032-308 10032-394 10032-770 10032-310 10032-312
Lab Coats, ESD Knit
Workwear
The ESD Knit Lab Coat is designed to provide excellent anti-static protection for electronics applications.
  • Excellent static dissipative properties
  • Lightweight, breathable fabric for added comfort when worn over regular clothes
  • Reinforced front facing and flat felled seams for improved durability
  • Three-position snap cuff for adjustability
The garments feature two lower pockets and one breast pocket. Each lab coat features a snap-front with reinforced facing and flat-felled seams for extra durability, and a three-position snap cuff.
Manufactured with a durable and breathable fabric, it offers a slight stretch for added comfort and ease of movement during asembly operations.
Ideal for use in micro-electronics assembly, microchip manufacturing, and semi-conductor production.
Ordering information: Available in blue only. Sizes 3X-Large and 4X-Large require extended lead time. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty