Would you like to log in again?
VWR® Micro Homogenizing Packages
Supplier: VWR
VWR®
VW-PK-01200P VW-PK-01200S VW-PK-01200D
10032-324KT 3001.54 USD InStock
10032-324 10032-326 10032-328
VWR® Micro Homogenizing Packages
Homogenizers Handheld Homogenizers
The VWR® Standard Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes.
  • Designed to mix all sample types
  • Quick and efficient processing
Volumes as small as 0.03 mL can be easily processed in a matter of seconds
The VWR® Deluxe Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes. The 5mm flat-bottom generator probe is designed for liquid and soft samples. A 7mm saw-tooth generator probe provides extra cutting action for all samples. The Deluxe Package offers full homogenizing capability for all types of samples within micro-tubes. From light mixing to processing tough tissue samples, this micro-homogenizing package can provide fast and complete breakdown results. Volumes as small as 0.03 mL can be easily processed in a matter of seconds.
The VWR® Premium Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes. The 5mm flat-bottom generator probe is designed for liquid and soft samples. A 7mm saw-tooth generator probe provides extra cutting action for all samples. Complete with stand assembly for hands-free homogenizing of all sample types within micro-tubes. From light mixing to processing tough tissue samples, the Premium micro-homogenizing package can provide fast and complete results without having to hold the homogenizer unit. Volumes as small as 0.03 mL can be easily processed in a matter of seconds.
The 5mm flat-bottom generator probe is designed for liquid and soft samples. Ideal for processing in micro-tubes, whether gently mixing, extracting RNA/DNA, or destroying the cell wall, this standard micro-homogenizing package can effectively and efficiently process virtually any small sample.
Certifications: CE marked.
Ordering information: Supplied with a two-year manufacturer's warranty. All VWR Homogenizing Packages include a carrying case with a foam insert to for protected storage. The Standard Micro-Homogenizing Package (10032-324) includes VWR® 200 Homogenizer unit and 5x75mm flat-bottom generator probe. The Deluxe Micro-Homogenizing Package (10032-326) includes VWR® 200 Homogenizer unit, 5x75mm flat-bottom generator probe, and 7x95mm saw-tooth generator probe. The Premium Micro-Homogenizing Package (10032-328) includes VWR® 200 Homogenizer unit, 5x75mm flat-bottom generator probe, 7x95mm saw-tooth generator probe, and stand assembly.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty