Would you like to log in again?
VWR® Universal Homogenizing Packages
Supplier: VWR
VWR®
VWPK01200UHD VWPK01200UHS
10032-332KT 4601.33 USD InStock
10032-332 10032-334
VWR® Universal Homogenizing Packages
Homogenizers Handheld Homogenizers
The VWR® Standard Universal Homogenizing Package is designed for use with all types of samples, even tough or stringy.
  • Designed for use with all types of samples, even tough or stringy
  • Allows flexibility
  • Fast and efficient operation
The package allows the flexibility of homogenizing within a wide range of tube sizes, from micro-tubes to large tubes (1.5–50 mL tubes)
The VWR® Deluxe Universal Homogenizing Package is also designed for use with all types of samples, even tough or stringy. This package allows the flexibility to process tubes ranging from 1.5–50 mL as well. The 7mm and 10mm saw-tooth generator probes provide fast and efficient operation. From soft to tough samples, both saw-tooth generator probes provide extra cutting action during homogenizing, ensuring fast and complete sample breakdown. The package is supplied complete with stand assembly for hands-free homogenizing within sizes ranging from micro-tubes to large tubes, without having to hold the homogenizer unit.
Both the 7 mm and 10 mm saw-tooth generator probes provide fast and efficient operation. From soft to tough samples, both saw-tooth generator probes provide extra cutting action during homogenizing, ensuring fast and complete sample breakdown.
Certifications: CE marked.
Ordering information: The VWR® Standard Universal Homogenizing Package (10032-332) includes VWR® 200 Homogenizer unit, 7x95mm saw-tooth generator probe, and 10x115mm saw-tooth generator probe. The VWR® Deluxe Universal Homogenizing Package (10032-334) includes VWR® 200 Homogenizer unit, 7x95mm saw-tooth generator probe, 10x115mm saw-tooth generator probe, and stand assembly.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty