Would you like to log in again?
Bead Ruptor Pre-Filled Bead Tubes, 2 ml, Omni International
Supplier: Omni International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
19-620 19-622 19-621 19-624 19-628 19-623 19-627
10032-358PK 202.62 USD InStock
10032-358 75839-548 10032-756 75839-448 75839-446 10032-360 10032-362
Bead Ruptor Pre-Filled Bead Tubes, 2 ml, Omni International
Tubes Centrifuge Tubes
Bead beating is the most effective and verstile lysis method available.
  • Certified RNase- and DNase-free to ensure high analyte integrity
2 ml skirted polypropylene screw cap tube with O-Ring. Pre-filled with lysing matrix. For use on all Omni International Bead Ruptor platforms. Bead beating tubes are designed to withstand the high forces generated on the Bead Ruptor Bead Mill.
Omni International offers users the ultimate flexibility with a range of bead beating tubes and lysing matrices to support efficient homogenization of a wide range of samples. Bead Ruptor bead beating tubes are pre-filled with an optimized amount of lysing matrix to provide efficient, consistent, and high-yield sample homogenates.
Ordering information: Unit includes 50 bead beating tubes pre-filled with lysing matrix.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty