Would you like to log in again?
Target DP™ 9-425 Screw-Thread Vial Caps, Thermo Scientific
Supplier: Thermo Scientific
Target DP™
60180-670 C5000-51A C5000-51Y C5000-51G C5000-51P 60180-669 C5000-52AE 60180-672 C5000-51R
97054-204PK 83.07 USD InStock
97054-204 89500-010 97054-202 10033-154 97054-208 89500-008 89500-006 89500-004 89500-012
Target DP™ 9-425 Screw-Thread Vial Caps, Thermo Scientific
Closures Chromatography Vial Closures
The Target DP 9 mm closures are designed to help chromatographers achieve more reliable and efficient sample analysis.
  • Septa push through nearly eliminated due to improved interior geometry
  • Increased sealing capability
  • AVCS provides the freedom to select the best septum for your instrument and applications
  • Cost efficient alternative to current choices for resolving push through (bonded septa)
  • Optimized ergonomics, fine texturing and evenly spaced ribbing for superior handling even with poorly fitting gloves
  • Rough edges have been eliminated making it easier and more comfortable to use while improving autosampler operation
Designed to accommodate nearly all chromatography autosamplers fitted for 12×32 cm vial trays, the C5000 series closure effectively eliminates septa push-through and significantly enhances productivity and flexibility in the laboratory. The C5000 series closures are fully compatible with all of the C4000 series Target DP 9 mm vials. With just one turn, they offer easy-on, easy-off convenience, and are a cost effective alternative to caps with bonded septa.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty