Would you like to log in again?
Lab Sinks, Self Rimming, Two Bowl, V-2 Series, Eagle MHC
Supplier: Eagle MHC
SR18A2V2 SR16B2V2 SR24B2V2 SR16C2V2 SR22A2V2 SR14D2V2
10033-256EA 3200.84 USD InStock
10033-256 10033-254 10033-252 10033-284 10033-282 10033-280
Lab Sinks, Self Rimming, Two Bowl, V-2 Series, Eagle MHC
Furniture Casework
Heavy gauge type 304 series stainless steel coved bowls with large radius.
  • Sink can be mounted on top or bottom of countertop
  • Two compartment
Basket drain of 1½" diameter in center of bowl. Features 1⅜" hole for mixing faucet and 1¼" hole for eye/drench hose.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty