Would you like to log in again?
Lab Sinks, Self Rimming, Three Bowl, V-3 Series, Eagle MHC
Supplier: Eagle MHC
SR16C3V3 SR14D3V3 SR16B3V3 SR18A3V3
10033-388EA 5161.79 USD InStock
10033-388 10033-386 10033-384 10033-382
Lab Sinks, Self Rimming, Three Bowl, V-3 Series, Eagle MHC
Furniture Casework
Heavy gauge type 304 series stainless steel coved bowls with large radius.
  • Three compartment
  • 1½" Ø basket drain in center of bowl
Sink can be mounted on top or bottom of countertop. Features ¾" hole for DI faucet, 1³/₈" hole for mixing faucet and 1¼" hole for eye/drench hose.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty