Would you like to log in again?
FLUOR DE LYS® SIRT5 fluorometric drug discovery assay kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BML-AK513-0001
10033-576EA 896.9 USD InStock
10033-576
FLUOR DE LYS® SIRT5 fluorometric drug discovery assay kit, Enzo Life Sciences
Assays Protein Assays
The SIRT5 Fluorometric Drug Discovery Kit is a sensitive fluorometric assay is designed to measure the lysyl desuccinylase activity of the recombinant human SIRT5.
  • Easy to use kit (two-step) for screening SIRT5 inhibitors/activators; includes enough active enzyme for entire plate
  • Suitable for high-throughput screening (Z’-factors >0.73)
  • Optimal, specific SIRT5 substrate means low enzyme concentration, making ‘hit’ validation easy
The SIRT5 Fluorometric Drug Discovery Kit contains an AMC (‘blue’) fluorophore that uses commonly-used wavelengths.
Delivery information: Kit contains SIRT5 (Sirtuin 5) (human, rec.), FLUOR DE LYS®-Succinyl, desuccinylase substrate, FLUOR DE LYS® Developer Concentrate (20X), NAD+ (Sirtuin Substrate), Nicotinamide (Sirtuin Inhibitor), Suramin sodium (Sirtuin Inhibitor), FLUOR DE LYS® Desuccinylated Standard, Sirtuin Assay Buffer, and half volume 96-well black NBS™ microplate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty