Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
iMiniPlus® Dry Ice Loggers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
MP-OE-N-8-L MP-ST-N-8-L
10033-818EA 134.17 USD InStock
10033-818 97017-230
iMiniPlus® Dry Ice Loggers, Cryopak Verification Technologies
Data Loggers
The iMiniPlus® Dry Ice logger can be programmed to measure extreme temperatures within the range of -100°C to 40°C (-148°F to 104°F)
  • Start and stop buttons on the face of the unitx
  • Bookmarking capabilities
  • Large LCD display with multifunctional viewing
  • Large memory capacity up to 8040 readings
  • Polycarbonate housing
Devices have a 1 m cable that connects the stainless steel temperature probe to the data logger. Available as a single-trip or multi-use unit.
It is the ideal solution for ensuring safe transport and storage of perishables held in dry ice.
The data logger box must not be inserted inside the extreme cold environment for risk of stopping the LCD screen from functioning, but particularly of severely damaging the iMiniPlus Dry Ice battery. The probe must therefore be inserted in the environment to be monitored and the box left outside it.
Certifications: ISO9001:2008, CE, RoHS, EN12830.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty