Would you like to log in again?
iMiniPlus PDF® Temperature Logger, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
MU-IN-D-16-L MU-OE-D-16-L MU-ST-D-8-L
10033-826EA 205.4 USD InStock
10033-826 10033-824 10033-822
iMiniPlus PDF® Temperature Logger, Cryopak Verification Technologies
Data Loggers
The iMiniPlus®, provided by Cryopak Verification Technologies, is an extension to the iMini® line
  • Two years warranty
  • The multifunctional LCD screen then provides several levels of information:
  • The highest temperature, the lowest temperature and the average temperature recorded
  • Any alarms triggered (high and/or low)
  • The time spent outside alarm ranges
  • Battery depletion
The iMiniPlus PDF® data logger uses a USB mini B to USB type A cable (supplied with PDF logger), or small USB adaptor for computer communications. It is a true plug and play device. When the logger is connected, the computer automatically recognizes the logger as a Mass Storage Device and a PDF report can be viewed with two other binary files. Users must allow sufficient time for installation (5 to 30 seconds).
Every data logger is supplied with a simple USB to USB cable for PC connection and travels with the logger in the consignment. Report generation includes a summary of transport condition, time spent out of specification, and trip statistics. Additionally, the report provides date, time, and temperature of all readings plus a graph of the temperature of the trip. The PDF report can be validated via Console software by emailing the PDF report and binary files to the supplier, thus meeting the requirements for 21 CFR Part11. All units offer an IP rating of IP65.
This data logger will generate a comprehensive PDF report, making it the ideal logger for shipments in distant locations where proprietary software is not available.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty